Learn More
Sino Biological Rhesus IL6ST Protein 200ug (Sino Biological)
Supplier:  Sino Biological 90275C02H200
Encompass_Preferred
A DNA sequence encoding the rhesus IL6ST (NP_001252920.1) (Met1-Ile618) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3847