Learn More
Sino Biological Rhesus LAMP3 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90247C08H100
Encompass_Preferred
A DNA sequence encoding the rhesus LAMP3 (Q8MJJ2) (Lys28-Thr381) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1697