Learn More
Sino Biological Anti-LAYN Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90160T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus Layilin / LAYN (Catalog#90160-C08H; XP_001105766.1; Met1-Glu220). Layilin / LAYN specific IgG was purified by Rhesus Layilin / LAYN affinity chromatography.
Catalog No. 50-240-8624