Learn More
Sino Biological Recombinant Rhesus LIFR Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90210K08H100
Encompass_Preferred
A DNA sequence encoding the Rhesus LIFR (XM_015139921.2) (Met1-Ser833) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-8809