Learn More
ACROBiosystems Rhesus macaque CD5 100ug
Catalog No. 502535371
Encompass_Preferred
Rhesus macaque CD5 Protein His Tag (CD5-R52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Thr 382 (Accession # F6V5Q9).
Catalog No. 50-253-5371 Supplier ACROBiosystems Supplier No. CD5R52H3100UG
Rhesus macaque CD5 Protein His Tag (CD5-R52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Thr 382 (Accession # F6V5Q9).