Learn More
ACROBiosystems ACROBiosystems Rhesus macaque DLL3 Protein, His Tag
Catalog No. 502106878
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque DLL3, His Tag (DL3-R52H4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 490 (Accession # F6TFV4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.2 kDa.
Catalog No. 50-210-6878 Supplier ACROBiosystems Supplier No. DL3R52H41MG
Recombinant Protein;1MG;Rhesus macaque DLL3, His Tag (DL3-R52H4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 490 (Accession # F6TFV4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.2 kDa.