Learn More
ACROBiosystems Rhesus macaque EpCAM 25ug
Catalog No. 502535090
Encompass_Preferred
Biotinylated Rhesus macaque EpCAM / TROP1 Protein HisAvitag (EPM-R82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # Q1WER1).
Catalog No. 50-253-5090 Supplier ACROBiosystems Supplier No. EPMR82E325UG
Biotinylated Rhesus macaque EpCAM / TROP1 Protein HisAvitag (EPM-R82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # Q1WER1).