Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque GITR / TNFRSF18 Protein, Fc Tag;HEK293;100UG;GIR,AITR,GITR,TNFRSF18,CD357
Catalog No. 501138333
Encompass_Preferred
Recombinant Protein;Rhesus macaque GITR / TNFRSF18 Protein, Fc Tag;100UG;Rhesus macaque GITR, Fc Tag (GIR-C5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # F7DDV8).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.3 kDa. As a result of glycosylation, the protein migrates as 48 kDa under reducing (R) condition, and 85 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-113-8333 Supplier ACROBiosystems Supplier No. GIRC5255100UG
Recombinant Protein;Rhesus macaque GITR / TNFRSF18 Protein, Fc Tag;100UG;Rhesus macaque GITR, Fc Tag (GIR-C5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # F7DDV8).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.3 kDa. As a result of glycosylation, the protein migrates as 48 kDa under reducing (R) condition, and 85 kDa under non-reducing (NR) condition (SDS-PAGE).