Learn More
Raybiotech Inc Rhesus Monkey IFN-gamma ELISA Kit
Supplier:  Raybiotech Inc ELKIFNG1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 24 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Rhesus IFN-gamma.
Catalog No. 50-194-9879