Learn More
Raybiotech Inc Rhesus Monkey IGFBP-3 ELISA Kit
Supplier:  Raybiotech Inc ELKIGFBP31
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 1.02 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Rhesus Macaque IGFBP-3 and human IGFBP-3. Other species not determined.
Catalog No. 50-194-9881