Learn More
ACROBiosystems Rhesus macaque IL-13 R alpha 2 Protein, His Tag
Catalog No. 502055090
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque IL-13 R alpha 2, His Tag (IL2-R52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 26 - Ther 340 (Accession # XP_014983540.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.0 kDa.
Catalog No. 50-205-5090 Supplier ACROBiosystems Supplier No. IL2R52H51MG
Recombinant Protein;1MG;Rhesus macaque IL-13 R alpha 2, His Tag (IL2-R52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 26 - Ther 340 (Accession # XP_014983540.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.0 kDa.