Learn More
Raybiotech Inc Rhesus IL-1beta ELISA Kit
Supplier:  Raybiotech Inc ELKIL1BETA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants.. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rhesus IL-1beta ELISA Kit.
Catalog No. 50-194-9885