Learn More
Raybiotech Inc Rhesus Monkey IL-6 R ELISA Kit
Supplier:  Raybiotech Inc ELKIL6SR1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 12 pg/ml . Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Rhesus Monkey and human IL-6R. Other species not determined.
Catalog No. 50-194-9888