Learn More
Raybiotech Inc Rhesus Monkey IL-8 (CXCL8) ELISA Kit
Supplier:  Raybiotech Inc ELKIL81
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Rhesus Macaque Interleukin 8 (CXCL8)
Catalog No. 50-194-9889