Learn More
Bon Opus Biosciences Recombinant Rhesus Macaque LAIR1 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences C02D50UG
Encompass_Preferred
Recombinant Rhesus Macaque Leukocyte-Associated Immunoglobulin-Like Receptor 1 is produced by our Mammalian expression system and the target gene encoding Gln22-Try165 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# A0A0U3SBU1. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C02D
Catalog No. 50-208-0343