Learn More
ACROBiosystems ACROBiosystems Rhesus macaque PD-1 / PDCD1 Protein, His Tag (MALS verified)
Catalog No. 502107282
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque PD-1, His Tag (PD1-R52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.9 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque PD-1, His Tag (Cat. No. PD1-R52H3) at 2 μg/mL (100 μL/well) can bind nivolumab with a linear range of 0.1-3 ng/mL (QC tested).;Immobilized Biotinylated Human PD-L2, Fc,Avitag (Cat. No. PD2-H82F6) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Rhesus macaque PD-1, His Tag (Cat. No. PD1-R52H3) with a linear range of 0.01-0.313 μg/mL (Routinely tested).
Catalog No. 50-210-7282 Supplier ACROBiosystems Supplier No. PD1R52H31MG
Recombinant Protein;1MG;Rhesus macaque PD-1, His Tag (PD1-R52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.9 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque PD-1, His Tag (Cat. No. PD1-R52H3) at 2 μg/mL (100 μL/well) can bind nivolumab with a linear range of 0.1-3 ng/mL (QC tested).;Immobilized Biotinylated Human PD-L2, Fc,Avitag (Cat. No. PD2-H82F6) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Rhesus macaque PD-1, His Tag (Cat. No. PD1-R52H3) with a linear range of 0.01-0.313 μg/mL (Routinely tested).