Learn More
Raybiotech Inc Rhesus Monkey Platelet Factor 4 (CXCL4) ELISA Kit
Supplier:  Raybiotech Inc ELKPF41
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.041 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Rhesus Macaque PF-4 and human PF-4. Other species not determined
Catalog No. 50-194-9894