Learn More
Raybiotech Inc Rhesus Monkey TNF alpha ELISA Kit
Supplier:  Raybiotech Inc ELKTNFA1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 40 pg/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair shows have approximately 15% cross-reactivity with the rrTNF-alpha and rmTNF-alpha.
Catalog No. 50-194-9896