Learn More
ACROBiosystems Rhesus macaque TROP-2 / TACSTD2 Protein, His Tag (MALS verified)
Catalog No. 502054909
Encompass_Preferred
Recombinant Protein;100UG;Rhesus macaque TROP-2, His Tag (TR2-R52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 274 (Accession # XP_001114599.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.4 kDa. The protein migrates as 38-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4909 Supplier ACROBiosystems Supplier No. TR2R52H3100UG
Recombinant Protein;100UG;Rhesus macaque TROP-2, His Tag (TR2-R52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 274 (Accession # XP_001114599.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.4 kDa. The protein migrates as 38-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.