Learn More
Raybiotech Inc Rhesus Monkey BDNF ELISA Kit
Supplier:  Raybiotech Inc ELKBDNF1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 2.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rhesus Monkey BDNF.
Catalog No. 50-194-9868