Learn More
Raybiotech Inc Rhesus Monkey CD14 ELISA Kit
Supplier:  Raybiotech Inc ELKCD141
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rhesus Monkey CD14.
Catalog No. 50-194-9871