Learn More
Lonza Walkersville Rhesus Monkey Heps Pleateable
Catalog No. 5000496319
Encompass
All our plateable cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are tested for healthy morphology lasting 72 hours in monolayer sandwich culture
Catalog No. 50-004-96319 Supplier Lonza Walkersville Supplier No. RHCP01
All our plateable cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are tested for healthy morphology lasting 72 hours in monolayer sandwich culture