Learn More
Lonza Walkersville Rhesus Monkey Heps Suspension
Catalog No. 5000496320
Encompass
All our suspension cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are not suitable for applications requiring adherant cells
Catalog No. 50-004-96320 Supplier Lonza Walkersville Supplier No. RHCS01
All our suspension cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are not suitable for applications requiring adherant cells