Learn More
iQ Biosciences Rhesus Monkey PBMCs, No Immunophenotyping, 10M cells/vial
Catalog No. NC3591095
Encompass_Preferred
High-quality, high-viability (> 90%) rhesus monkey (or macaque) peripheral blood mononuclear cells (PBMCs) sourced responsibly from a vetted third party facility. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor - Donor(s) with a higher percentage of the cells of interest, i.e., B cells, CD14+ cells, CD4+ cells, CD8+ cells, or NK cells
Catalog No. NC3591095 Supplier iQ Biosciences Supplier No. IQBMNPB210NP
High-quality, high-viability (> 90%) rhesus monkey (or macaque) peripheral blood mononuclear cells (PBMCs) sourced responsibly from a vetted third party facility. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor - Donor(s) with a higher percentage of the cells of interest, i.e., B cells, CD14+ cells, CD4+ cells, CD8+ cells, or NK cells