Learn More
Sino Biological Anti-PD-1 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 90305MM35F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus PD-1/PDCD1 ( Catalog#90305-K08H; NP_001107830.1; Met1-Gln167) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1208