Learn More
Sino Biological Anti-PD-1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 90305MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus PD-1 / PDCD1 ( Catalog#90305-K08H; NP_001107830.1; Met1-Gln167). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2234