Learn More
Sino Biological Anti-PD-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90305T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus PD1/PDCD1/CD279 ( Catalog#90305-K08H; NP_001107830.1; Met1-Gln167). PD1/PDCD1/CD279 specific IgG was purified by Rhesus PD1/PDCD1/CD279 affinity chromatography.
Catalog No. 50-240-8681