Learn More
Sino Biological Rhesus PDCD1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90305K08H100
Encompass_Preferred
A DNA sequence encoding the rhesus PD1 (NP_001107830.1) (Met1-Gln167) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2159