Learn More
Sino Biological Rhesus PDCD1LG2 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 90249K02H1
Encompass_Preferred
A DNA sequence encoding the Rhesus PDCD1LG2 (A4GW30) (Met1-Pro219) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1762