Learn More
Sino Biological Rhesus PVR Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90005C08H50
Encompass_Preferred
A DNA sequence encoding the rhesus PVR (NP_001036851.1) extracellular domain (Met 1-Asn 343) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-237-2705