Learn More
Sino Biological Rhesus REG1B Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90163C02H100
Encompass_Preferred
A DNA sequence encoding the rhesus REG1B (NP_001181497.1) (Met1-Asn166) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3592