Learn More
Sino Biological Recombinant Rhesus P-Selectin/CD62P Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90170C08H100
Encompass_Preferred
A DNA sequence encoding the rhesus SELP (XP_001094728.1) (Met1-Ala771) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3823