Learn More
Sino Biological Anti-Semaphorin 4D Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90209T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus Semaphorin 4D / SEMA4D / CD100 (Catalog#90209-C08H; EHH24221.1; Met1-Arg734). Semaphorin 4D / SEMA4D / CD100 specific IgG was purified by Rhesus Semaphorin 4D / SEMA4D / CD100 affinity chromatography.
Catalog No. 50-255-2708