Learn More
Sino Biological Anti-SP-D/SFTPD Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90003RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus SFTPD / SP-D / SFTP4 (Catalog#90003-C08H; NP_001035283.1; Met1-Phe375). SFTPD / SP-D / SFTP4 specific IgG was purified by Rhesus SFTPD / SP-D / SFTP4 affinity chromatography.
Catalog No. 50-240-7857