Learn More
Sino Biological Recombinant Rhesus CD4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90274C08H1
Encompass_Preferred
A DNA sequence encoding the rhesus CD4 (EHH20435.1) (Met1-Trp390) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-222-3836