Learn More
Sino Biological Rhesus TNFRSF10D Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90100C02H100
Encompass_Preferred
A DNA sequence encoding the rhesus TNFRSF10D (XP_001107922.1) (Met1-Leu206) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3304