Learn More
Sino Biological Recombinant Cynomolgus, Rhesus BCMA Protein (AVI & Fc Tag), Biotinylated
Supplier:  Sino Biological 90103C35HB100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus / Rhesus TNFRSF17 (XP_001106892.1) (Met1-Ala53) was expressed with a c-terminal AVI tagged Fc region of human IgG1 tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2719