Learn More
Sino Biological Anti-TNFR-2 / CD120b Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90102T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus / Rhesus TNFR2 / CD120b / TNFRSF1B (Catalog#90102-C08H; XP_005544817.1/NP_001253134.1; Met1-Asp257). TNFR2 / CD120b / TNFRSF1B specific IgG was purified by Cynomolgus / Rhesus TNFR2 / CD120b / TNFRSF1B affinity chromatography.
Catalog No. 50-240-7882