Learn More
Sino Biological Rhesus TNFRSF4 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90846C02H50
Encompass_Preferred
A DNA sequence encoding the rhesus TNFRSF4 (XP_001090870.1) (Met1-Ala216) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3543