Learn More
Sino Biological Anti-VISTA/B7-H5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90801RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus B7-H5 / GI24 / VISTA ( Catalog#90801-K08H; NP_001177807.1; Met1-Ala194). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1215