Learn More
Cayman Chemical Rheum hotaoense AstrngIn 1mg
SDP
Catalog No. 502941802
Encompass_Preferred
A phenolic stilbene glucoside with antioxidant and antineoplastic activities; scavenges DPPH radicals (IC50 = 30.2 μM) in a cell-free assay and inhibits cupric ion-induced lipid peroxidation of human LDL (IC50 = 3 μM); inhibits development of DMBA-induced preneoplastic lesions in mouse mammary gland organ cultures by 68.8% at 10 UG/ml
Catalog No. 50-294-1802 Supplier Cayman Chemical Supplier No. 279791 MG
A phenolic stilbene glucoside with antioxidant and antineoplastic activities; scavenges DPPH radicals (IC50 = 30.2 μM) in a cell-free assay and inhibits cupric ion-induced lipid peroxidation of human LDL (IC50 = 3 μM); inhibits development of DMBA-induced preneoplastic lesions in mouse mammary gland organ cultures by 68.8% at 10 UG/ml