Learn More
Innovative Research Inc Single Donor Human Rheumatoid Arthritis Serum
Catalog No. 502036254
Encompass_Preferred
Single Donor Human Rheumatoid Arthritis Serum from Innovative Research is whole blood derived. Available volumes are variable, and range from 0.5ml to 1.0ml per vial (you can select your exact samples from our inventory, if desired). A detailed patient history and clinical information is recorded at the time of draw, and multiple samples and plasma/serum matched sets from the same donor are often available.
Catalog No. 50-203-6254 Supplier Innovative Research Inc Supplier No. ISERSRA
Single Donor Human Rheumatoid Arthritis Serum from Innovative Research is whole blood derived. Available volumes are variable, and range from 0.5ml to 1.0ml per vial (you can select your exact samples from our inventory, if desired). A detailed patient history and clinical information is recorded at the time of draw, and multiple samples and plasma/serum matched sets from the same donor are often available.