Learn More
Azzota Corp RHF-200T antifreeze refract
Catalog No. 5000887274
Encompass_Preferred
RHF-200T Battery/Antifreeze/Cleaning Fluid Refractometer With ATC Ethylene Glycol -60 C to 0 C Propylene Glycol -50 C to 0 C Battery 1 100-1 400kg/l
Catalog No. 50-008-87274 Supplier Azzota Corp Supplier No. RHF200T
RHF-200T Battery/Antifreeze/Cleaning Fluid Refractometer With ATC Ethylene Glycol -60 C to 0 C Propylene Glycol -50 C to 0 C Battery 1 100-1 400kg/l