Learn More
Creative Biomart Meningococcal polysaccharide Group Y Vaccine Antigen
Supplier:  Creative Biomart VAC0318
Encompass_Preferred
Meningococcal group Y polysaccharide is group-specific polysaccharide antigens extracted and purified from Neisseria meningitidis serogroup Y. N. meningitidis is a bacteria that causes endemic and epi
Catalog No. 50-208-5158