Learn More
Medchemexpress LLC RHI002-Me | 314261-66-0 | 98.5% | 373.49 | C18H19N3O2S2 | 25MG
SDP
Catalog No. 5000237631
Encompass_Preferred
RHI002-Me is a small-molecule derivative of RHI002 reported as an hRNaseH2 inhibitor for research use. It is supplied as a solid in multiple milligram quantities and as a ready-to-use 10 mM solution in DMSO, and is accompanied by analytical documents for identity and handling. Typical applications include biochemical assays and cellular studies investigating RNase H2-related pathways.
  • hRNaseH2 inhibitor suitable for biochemical and cell-based research.
  • Available as solid (5 mg-100 mg) and 10 mM in DMSO (1 mL).
  • High purity, typically 98.5% by HPLC.
  • Molecular weight 373.49 and formula C18H19N3O2S2.
  • Analytical documentation available: COA, SDS, H-NMR, RP-HPLC, MS, handling instructions.
Catalog No. 50-002-37631 Supplier Medchemexpress LLC Supplier No. HY12369225MG
RHI002-Me is a small-molecule derivative of RHI002 reported as an hRNaseH2 inhibitor for research use. It is supplied as a solid in multiple milligram quantities and as a ready-to-use 10 mM solution in DMSO, and is accompanied by analytical documents for identity and handling. Typical applications include biochemical assays and cellular studies investigating RNase H2-related pathways.
  • hRNaseH2 inhibitor suitable for biochemical and cell-based research.
  • Available as solid (5 mg-100 mg) and 10 mM in DMSO (1 mL).
  • High purity, typically 98.5% by HPLC.
  • Molecular weight 373.49 and formula C18H19N3O2S2.
  • Analytical documentation available: COA, SDS, H-NMR, RP-HPLC, MS, handling instructions.