Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Rhinovirus A80 Culture Fluid
SDP

Catalog No. 22158114 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-114 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-114 Supplier ZeptoMetrix Supplier No. 0810288CF
Add to Cart
Add to Cart

Intended for research, product development testing, or quality assurance testing

Rhinovirus consists of a non-enveloped capsid containing a positive-sense single-stranded RNA.

  • Propagated in the MRC-5 or HeLa cell line
  • Each lot is assayed for its tissue culture infective dose (TCID50) and sold with titers >105 U/mL

Specifications

Biosafety Level 2
Content And Storage -65°C or below
For Use With (Application) Nucleic acid/molecular testing, Limit of Detection (LOD) studies, Cross-reactivity studies, Other viral-based assays
Quantity 1 mL

Do not use in humans. For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.