Learn More
ATCC Rhizobium rhizogenes
Catalog No. 502383150
Encompass
Rhizobium rhizogenes strain [CIP 104786] is a whole-genome sequenced strain that produces isoprene. This bacterium has applications in agricultural research.
Catalog No. 50-238-3150 Supplier ATCC Supplier No. 15834
Rhizobium rhizogenes strain [CIP 104786] is a whole-genome sequenced strain that produces isoprene. This bacterium has applications in agricultural research.