Learn More
ATCC Rhizoctonia solani
Catalog No. 502383006
Encompass
Major potato pathogen that causes black scurf, root rot, and Rhizoctonia canker. This strain was isolated in 1926 from a potato in Colorado, United States. This product is provided as an ampoule containing viable cells (yeast cells, spores, or agar cubes with mycelia) suspended in cryoprotectant.
Catalog No. 50-238-3006 Supplier ATCC Supplier No. 10182
Major potato pathogen that causes black scurf, root rot, and Rhizoctonia canker. This strain was isolated in 1926 from a potato in Colorado, United States. This product is provided as an ampoule containing viable cells (yeast cells, spores, or agar cubes with mycelia) suspended in cryoprotectant.