Learn More
Aviva Systems Biology Corp RHO Antibody - C-terminal region (ARP59909_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59909_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHO. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human RHO. Alternate names: CSNBAD1, MGC138309, MGC138311, OPN2, RP4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-286-873