Learn More
Cell Signaling Technology Rho-GTPase Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502729449
Encompass
The Rho-GTPase Antibody Sampler Kit provides a comprehensive set of reagents designed to investigate key aspects of cell behavior, including cell migration, adhesion, proliferation, and differentiation. The kit contains high-quality primary antibodies to target members of the Rho-GTPase family, along with a secondary antibody to facilitate detection in Western blot experiments.
  • Includes primary antibodies for RhoA, RhoB, RhoC, Rac1, Rac2, Rac3, and Cdc42.
  • Supplied with an HRP-linked secondary antibody for Western blot analysis.
  • Validated for use in Western blotting, immunofluorescence, and immunohistochemistry.
  • Sufficient reagents to perform two Western blot experiments per primary antibody.
  • Supplied in a storage buffer containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
Catalog No. 50-272-9449 Supplier Cell Signaling Technology Supplier No. 9968T
The Rho-GTPase Antibody Sampler Kit provides a comprehensive set of reagents designed to investigate key aspects of cell behavior, including cell migration, adhesion, proliferation, and differentiation. The kit contains high-quality primary antibodies to target members of the Rho-GTPase family, along with a secondary antibody to facilitate detection in Western blot experiments.
  • Includes primary antibodies for RhoA, RhoB, RhoC, Rac1, Rac2, Rac3, and Cdc42.
  • Supplied with an HRP-linked secondary antibody for Western blot analysis.
  • Validated for use in Western blotting, immunofluorescence, and immunohistochemistry.
  • Sufficient reagents to perform two Western blot experiments per primary antibody.
  • Supplied in a storage buffer containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.